Learn More
Aviva Systems Biology Corp Eras Antibody - N-terminal region (ARP55793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eras. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: HRAS2, HRasp, Ha-Ras2, ecat5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-082