Learn More
Apexbio Technology LLC Erastin, 571203-78-6, MFCD09837984, 5 mg
SDP
Supplier:  Apexbio Technology LLC B152410
Encompass_Preferred
MF: C30H31ClN4O4, MW: 547.04, Solubility: 10.92mg/mL in DMSO with gentle warming. Erastin is a cell-permeable ferroptosis activatior and an antitumor agent that is selective for cell expressing oncogene RAS. Other sizes are available. Please inqury us for quote.
Catalog No. NC1138209