Learn More
Medchemexpress LLC HY-15763 1mg Medchemexpress, Erastin CAS:571203-78-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY157631MG
Encompass_Preferred
Medchemexpress, HY-15763 1mg Erastin CAS:571203-78-6 Erastin is a ferroptosis inducer. Erastin binds and inhibits voltage-dependent anion channels (VDAC2/VDAC3). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9167