Learn More
Selleck Chemical LLC Erastin
SDP
Supplier:  Selleck Chemical LLC S72422MG
Encompass_Preferred
Erastin is a ferroptosis activator by acting on mitochondrial VDAC exhibiting selectivity for tumor cells bearing oncogenic RAS Solutions are unstable and should be fresh-prepared
Catalog No. 50-000-02842