Learn More
Apexbio Technology LLC Erastin, 5mg. Cas: 571203-78-6 MFCD: MFCD09837984. cell-permeable ferroptosis activator and antitumor agent
SDP
Supplier:  Apexbio Technology LLC B15245
Encompass_Preferred
Erastin is a ferroptosis activator by acting on mitochondrial VDAC, exhibiting selectivity for tumor cells bearing oncogenic RAS. 571203-78-6. MFCD09837984. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-3395