Learn More
Medchemexpress LLC Erastin2 | 1695533-44-8 | 98.6% | 623.14 | 1 ML
SDP
Catalog No. 5000348196
Encompass_Preferred
Erastin2 is an analog of Erastin and acts as a ferroptosis inducer. It is a potent, selective inhibitor of the system xc(-) cystine/glutamate transporter.
  • Ferroptosis inducer
  • Potent, selective inhibitor of system xc(-) cystine/glutamate transporter
  • Available as a 10 mM solution in DMSO
  • For research use only
Catalog No. 50-003-48196 Supplier Medchemexpress LLC Supplier No. HY13908710MM/1ML
Erastin2 is an analog of Erastin and acts as a ferroptosis inducer. It is a potent, selective inhibitor of the system xc(-) cystine/glutamate transporter.
  • Ferroptosis inducer
  • Potent, selective inhibitor of system xc(-) cystine/glutamate transporter
  • Available as a 10 mM solution in DMSO
  • For research use only