Learn More
Selleck Chemical LLC Erastin2-E1874-5MG
SDP
Supplier:  Selleck Chemical LLC E18745MG
Encompass_Preferred
Erastin2 is an inducer of ferroptosis It also selectively inhibits xc(-) cystine/glutamate transporter It induces ferroptosis in HT-1080 T98G and A549 cells with the potential to treat neurodegenerative diseases
Catalog No. 50-313-4457