Learn More
Medchemexpress LLC Eravacycline-d4 (dihydrochloride) | 100.0% | 635.50 | 500 UG
SDP
Catalog No. 5000390633
Encompass_Preferred
Eravacycline-d4 (TP-434-d4) dihydrochloride is a deuterium labeled Eravacycline, functioning as a potent and broad-spectrum antibacterial agent. It is designed for research use and can be utilized as a tracer or an internal standard.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-90633 Supplier Medchemexpress LLC Supplier No. HY16980S500UG
Eravacycline-d4 (TP-434-d4) dihydrochloride is a deuterium labeled Eravacycline, functioning as a potent and broad-spectrum antibacterial agent. It is designed for research use and can be utilized as a tracer or an internal standard.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.