Learn More
Medchemexpress LLC Eravacycline-d4 dihydrochloride | 1207283-85-9 | 99.98% | 635.50 | 1 MG
SDP
Catalog No. 5000390567
Encompass_Preferred
Eravacycline-d4 (TP-434-d4) dihydrochloride is deuterium labeled Eravacycline. Eravacycline is a potent and broad-spectrum antibacterial agent. For research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90567 Supplier Medchemexpress LLC Supplier No. HY16980S1MG
Eravacycline-d4 (TP-434-d4) dihydrochloride is deuterium labeled Eravacycline. Eravacycline is a potent and broad-spectrum antibacterial agent. For research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.