Learn More
Medchemexpress LLC Eravacycline-d4 dihydrochloride | 99.98% | 635.50 | 5 MG
SDP
Catalog No. 5000390480
Encompass_Preferred
Eravacycline-d4 (TP-434-d4) dihydrochloride is deuterium labeled Eravacycline. Eravacycline is a potent and broad-spectrum antibacterial agent. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-90480 Supplier Medchemexpress LLC Supplier No. HY16980S5MG
Eravacycline-d4 (TP-434-d4) dihydrochloride is deuterium labeled Eravacycline. Eravacycline is a potent and broad-spectrum antibacterial agent. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.