Learn More
TARGETMOL CHEMICALS INC ERAVACYCLINE DIHYDROCHLO 5MG
SDP
Catalog No. 502581549
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Eravacycline dihydrochloride (TP-434-046)is a potent and broad-spectrum antibacterial agent against six E. coli (MICs 0.125-0.25 mg/L). purity: 95%
Catalog No. 50-258-1549 Supplier TARGETMOL CHEMICALS INC Supplier No. T11227L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Eravacycline dihydrochloride (TP-434-046)is a potent and broad-spectrum antibacterial agent against six E. coli (MICs 0.125-0.25 mg/L). purity: 95%