Learn More
ERB™ Boas™ Protective Eyewear
Description
- Ultra-lightweight
- Wraparound design fits snugly for total optical protection
- Features a 9.75 in.-base spherical polycarbonate lens
- Stainless-steel hinge screws
Specifications
Specifications
| Certifications/Compliance | ANSI/ISEA Z87.1 - Impact |
| Lens Tint | Clear |
| Frame Color | Clear |
| No. per Pack | 12/Pk. |
| Lens Material | Polycarbonate |