Learn More
ERB™ Omega II™ Suspension Hard Hats with Mega Ratchets
SDP
Catalog No. 19811298
Change view
Click to view available options
Color: yellow; With reflective strap
Catalog No.
19-811-298 Color: yellow; With reflective strap
1 options
Catalog No. 19-811-298 Supplier ERB™ Supplier No. 19952/GADOT/REFSTRIP
Safety helmet with mega ratchets
  • Accommodates chin strap
  • Accessory slots accommodate a wide variety of eye, face and hearing products
  • Certified when headband is installed in reverse position
  • Goggle retaining slot on suspension
  • Moisture wicking brow pad is removable, washable and replaceable
  • Meets the low temperature (LT) option of the Z89.1 standard
Specifications
Certifications/Compliance ANSI/ISEA Z89.1, Type 1, Class C, E and G requirements; SEI certified; OSHA
Color Yellow
Material Polyethylene
No. of Mounting Points 6
Size (Metric) 165.1 to 203.2 mm
Suspension Type Omega II™ Cap and 6-point ratchet suspension with attached brow pad