Learn More
ERB™ Omega II™ 6-Point Nylon Suspension Cap
SDP
Catalog No. 19811296
Change view
Click to view available options
Color: White
Catalog No.
19-811-296 Color: White
1 options
Catalog No. 19-811-296 Supplier ERB™ Supplier No. 19131
  • Accommodates chin strap
  • Accessory slots accommodate a wide variety of eye, face and hearing products
  • Certified when headband is installed in reverse position
  • Goggle retaining slot on suspension
  • Moisture-wicking brow pad is removable, washable and replaceable
  • Meets the low temperature (LT) option of Z89.1 standard
Specifications
Certifications/Compliance ANSI/ISEA Z89.1, Type 1, Class C, E and G requirements; SEI certified; OSHA
Color White
Includes Omega II™ cap and 6-point slide-lock suspension with attached brow pad
Material HDPE
Quantity 12/Pk.
Size 165.1 to 203.2 mm
Suspension Type 6-Point slide-lock suspension