Learn More
ERB™ Omega II™ Cap Replacement Suspension
SDP
Catalog No. 19811297
Change view
Click to view available options
Quick-Slide standard replacement suspension
Catalog No.
19-811-297 Quick-Slide standard replacement suspension
1 options
Catalog No. 19-811-297 Supplier ERB™ Supplier No. 19141
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
  • Quick-Slide Standard Replacement Suspension
  • Fits Omega II and Omega II Full Brim Safety Helmets 
  • Comfortable impact-absorbing 6-point woven nylon suspension
Specifications
Certifications/Compliance ANSI or OSHA
Color Gray
Includes I-85 six-point slide-lock suspension
Material Woven Nylon
Size 165.1 to 203.2 mm
Size (Metric) 165.1 to 203.2 mm
Suspension Type 6-point slide-lock suspension