Learn More
OriGene ErbB 3 (ERBB3) Human Recombinant Protein, High Purity > 80%, Facilitates Cancer Research, 1 mg
SDP
Catalog No. NC3729823
Encompass_Preferred
The ErbB 3 (ERBB3) Human Recombinant Protein is an essential reagent for research in cell biology and oncology. It plays a critical role in the epidermal growth factor receptor (EGFR) signaling pathways, particularly through heterodimerization with kinase-active partners. This recombinant protein enhances detection and purification with its C-DDK/His tag and is over 80% pure. Its unique characteristics set it apart for studies related to cancer, making it a valuable tool for researchers.
Catalog No. NC3729823 Supplier OriGene Supplier No. TP309954L1MG
Add to Cart
Add to Cart
Request a Quote
The ErbB 3 (ERBB3) Human Recombinant Protein is an essential reagent for research in cell biology and oncology. It plays a critical role in the epidermal growth factor receptor (EGFR) signaling pathways, particularly through heterodimerization with kinase-active partners. This recombinant protein enhances detection and purification with its C-DDK/His tag and is over 80% pure. Its unique characteristics set it apart for studies related to cancer, making it a valuable tool for researchers.