Learn More
Description
Hybridization signals of digoxigenin-labeled polynucleotides appear as dark green colored distinct dots (ERBB2 gene region), and dinitrophenyl-labeled polynucleotides appear as bright red colored distinct dots (CEN 17).
Normal situation: In interphases of normal cells or cells without an amplification involving the ERBB2 gene locus, two green signals and two red signals appear.
Aberrant situation: In cells with amplification of the ERBB2 gene locus, an increased number of gene specific signals or signal clusters are visible. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Specifications
| Content And Storage | The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. |
| For Use With (Application) | CISH-P |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.