Learn More
Description
Hybridization signals of digoxigenin-labeled polynucleotides appear as brown- to dark brown colored distinct dots (ERBB2 gene region).
Normal situation: In interphases of normal cells or cells without an amplification involving the ERBB2 gene locus, two distinct dot-shaped brown signals per nuclei appear.
Aberrant situation: In a cell with amplification of the ERBB2 gene locus or polysomy of chromosome 17, multiple copies of the brown signal or brown signal clusters will be observed. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Specifications
| Content And Storage | The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. |
| For Use With (Application) | CISH-P |
| Includes | Reagent Provided: 1. Digoxigenin-labeled polynucleotides targeting sequences mapping in 17q12* (chr17:37,725,661-37,973,541) harboring the ERBB2 gene 2. Formamide based hybridization buffer *according to Human Genome Assembly GRCh37/hg19 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.