Learn More
ACROBiosystems ACROBiosystems Mouse ErbB3 / Her3 Protein, His Tag
Catalog No. 501956398
Encompass_Preferred
Recombinant Protein;100UG;Mouse ErbB3, His Tag (ER3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - His 641 (Accession # Q61526-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.4 kDa.
Catalog No. 50-195-6398 Supplier ACROBiosystems Supplier No. ER3M52H5100UG
Recombinant Protein;100UG;Mouse ErbB3, His Tag (ER3-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - His 641 (Accession # Q61526-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.4 kDa.