Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human ErbB3 / Her3 Protein, His Tag
Supplier:  ACROBiosystems ER3HF2H525UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1). It is the FITC labeled form of Human ErbB3, His Tag (ER3-H5223).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6729