Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ErbB3 / Her3 Protein, His,Avitag™
Supplier: ACROBiosystems ER3H82E625UG
Recombinant Protein;25UG;Biotinylated Human ErbB3, His,Avitag (ER3-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 72.4 kDa. The protein migrates as 95-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NRG1-beat 1 Protein at 2 μg/mL (100 μL/well) can bind Biotinylated Human ErbB3, His,Avitag (Cat. No. ER3-H82E6) with a linear range of 0.039-0.625 μg/mL (QC tested).