Learn More
ACROBiosystems ACROBiosystems Human ErbB3 / Her3 Protein, Mouse IgG2a Fc Tag
Catalog No. 501956671
Encompass_Preferred
Recombinant Protein;100UG;Human ErbB3, Mouse IgG2a Fc Tag (ER3-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 95.5 kDa. The protein migrates as 115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NRG1-beta1 Protein (ECD, Fc Tag) at 2 μg/mL (100 μL/well) can bind Human ErbB3, Mouse IgG2a Fc Tag (Cat. No. ER3-H5257) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-195-6671 Supplier ACROBiosystems Supplier No. ER3H5257100UG
Recombinant Protein;100UG;Human ErbB3, Mouse IgG2a Fc Tag (ER3-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 95.5 kDa. The protein migrates as 115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human NRG1-beta1 Protein (ECD, Fc Tag) at 2 μg/mL (100 μL/well) can bind Human ErbB3, Mouse IgG2a Fc Tag (Cat. No. ER3-H5257) with a linear range of 0.3-2 ng/mL (QC tested).