Learn More
ACROBiosystems ACROBiosystems Human ErbB3 / Her3 Protein, Strep Tag
Catalog No. 501957469
Encompass_Preferred
Recombinant Protein;1MG;Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 71.7 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ErbB3, Strep Tag (Cat. No. ER3-H5288) at 2 μg/mL (100 μL/well) can bind Human NRG1-beta 1 Protein with a linear range of 1-10 ng/mL (Routinely tested).
Catalog No. 50-195-7469 Supplier ACROBiosystems Supplier No. ER3H52881MG
Recombinant Protein;1MG;Human ErbB3, Strep Tag (ER3-H5288) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 71.7 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ErbB3, Strep Tag (Cat. No. ER3-H5288) at 2 μg/mL (100 μL/well) can bind Human NRG1-beta 1 Protein with a linear range of 1-10 ng/mL (Routinely tested).