Learn More
ACROBiosystems Recombinant Protein;Human ErbB3 / Her3 Protein, Fc Tag;HEK293;200UG;ER3,ERBB3,ErbB-3,HER3,HER-3,LCCS2,MDA-BF-1,MGC88033,c-erbB-3,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3
Supplier:  ACROBiosystems ER3H5259200UG
Encompass_Preferred
Recombinant Protein;Human ErbB3 / Her3 Protein, Fc Tag;200UG;Human ErbB3, Fc Tag (ER3-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # NP_001973.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95.4 kDa. The protein migrates as 130-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7570