Learn More
Eurofins Discoverx ErbB4, active, 10 µg. (14-569)
Supplier:  Eurofins Discoverx 14569
Encompass
N-terminal 6xHis-tagged recombinant, human ErbB4 amino acids 706-991. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8983