Learn More
Santa Cruz Biotechnology ErbB4/HER4 Antibody (3F168) 200 µg/ml
Catalog No. NC3243643
Encompass
ErbB4/HER4 Antibody (3F168) is a mouse monoclonal IgG2b κ, cited in 1 publications, provided at 200 µg/mlraised against amino acids 1249-1264 of ErbB-4 of human originrecommended for detection of ErbB-4 of mouse, rat and human origin by WB, IP, IF and IHC(P)
Catalog No. NC3243643 Supplier Santa Cruz Biotechnology Supplier No. SC71070
ErbB4/HER4 Antibody (3F168) is a mouse monoclonal IgG2b κ, cited in 1 publications, provided at 200 µg/mlraised against amino acids 1249-1264 of ErbB-4 of human originrecommended for detection of ErbB-4 of mouse, rat and human origin by WB, IP, IF and IHC(P)