Learn More
ACROBiosystems ACROBiosystems Human ErbB4 / Her4 Protein, His Tag
Supplier:  ACROBiosystems ER4H5221100UG
Encompass_Preferred
Recombinant Protein;100UG;Human ErbB4, His Tag (ER4-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # NP_005226.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.7 kDa. The protein migrates as 95-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ErbB4, His Tag (Cat. No. ER4-H5221) at 2 μg/mL (100 μL/well) can bind Human NRG1-beta 1 Protein with a linear range of 1-10 ng/mL (Routinely tested).
Catalog No. 50-195-7467