Learn More
ACROBiosystems Recombinant Protein;Human ErbB4 / Her4 Protein, Fc Tag;HEK293;100UG;ER4,HER4,ErbB4,MGC138404,p180erbB4,HER-4,ErbB-4
Supplier:  ACROBiosystems ER4H5251100UG
Encompass_Preferred
Recombinant Protein;Human ErbB4 / Her4 Protein, Fc Tag;100UG;Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # NP_001036064.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96 kDa. The protein migrates as 122-132 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7935