Learn More
ACROBiosystems Recombinant Protein;Human ErbB4 / Her4 Protein, Fc Tag;HEK293;1MG;ER4,HER4,ErbB4,MGC138404,p180erbB4,HER-4,ErbB-4
Catalog No. 501657936
Encompass_Preferred
Recombinant Protein;Human ErbB4 / Her4 Protein, Fc Tag;1MG;Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # NP_001036064.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96 kDa. The protein migrates as 122-132 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7936 Supplier ACROBiosystems Supplier No. ER4H52511MG
Recombinant Protein;Human ErbB4 / Her4 Protein, Fc Tag;1MG;Human ErbB4, Fc Tag (ER4-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Pro 651 (Accession # NP_001036064.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96 kDa. The protein migrates as 122-132 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.