Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2100MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: ESR2/686. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a. Positive Control: MCF-7 Cells. Ovarian, Breast, Bladder, Gastric or Salivary carcinoma. Cellular Staining: Predominantly nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: C-terminus fragment of recombinant human estrogen receptor beta protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 53-59kDa.Chromosome: 14q23.2. Gene Symbol: ESR2. Gene ID: 2100. Ensemble: ENSG00000140009. Uniprot: NP_001428.
Catalog No. 50-188-4675