Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Catalog No. AA4048122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4048122 100 g
AA4048106 5 g
AA4048114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4048122 Supplier Thermo Scientific Chemicals Supplier No. 04048122
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15280-57-6

Erbium(III) acetate tetrahydrate participates in the synthesis and determination of crystal structures and stereochemical properties of lanthanide(III) complexes with ethylenediamine-N,N,N?,N?-tetraacetate. Participates in simple one-pot single-step synthesis of rare earth doped spherical mesoporous metal oxide nanospheres with upconversion emission ability in supercritical methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-57-6
MDL Number MFCD00013034

Specifications

Physical Form Crystalline Aggregates
Quantity 100 g
Linear Formula Er(OOCCH3)3·4H2O
Solubility Information Soluble in water.
Formula Weight 416.45 (344.39 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 2.114 g/mL
Assay (REO)
Chemical Name or Material Erbium(III) acetate tetrahydrate
Show More Show Less
EINECSNumber 247-067-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.