Learn More
Strem, An Ascensus Company CAS# 10138-41-7. 5g. Erbium(III) chloride, anhydrous (99.9%-Er) (REO). MFCD00010989
SDP
Catalog No. 5090116162
Encompass
CAS# 10138-41-7. 5g. Erbium(III) chloride, anhydrous (99.9%-Er) (REO). MFCD00010989. Molecular Weight: 273.64. Molecular Formula: ErCl3. Color/form: pink pwdr. Strem# 93-6803. http://www.strem.com/catalog/v/93-6803/
Catalog No. 50-901-16162 Supplier Strem, An Ascensus Company Supplier No. 9368035G
CAS# 10138-41-7. 5g. Erbium(III) chloride, anhydrous (99.9%-Er) (REO). MFCD00010989. Molecular Weight: 273.64. Molecular Formula: ErCl3. Color/form: pink pwdr. Strem# 93-6803. http://www.strem.com/catalog/v/93-6803/