Learn More
Strem, An Ascensus Company CAS# 10025-75-9. 10g. Erbium(III) chloride hydrate (99.999%-Er) (REO) PURATREM. MFCD00149692
SDP
Catalog No. 5090112860
Encompass
CAS# 10025-75-9. 10g. Erbium(III) chloride hydrate (99.999%-Er) (REO) PURATREM. MFCD00149692. Molecular Weight: 273.64. Molecular Formula: ErCl3∙XH2O. Color/form: pink xtl. Strem# 68-6804. http://www.strem.com/catalog/v/68-6804/
Catalog No. 50-901-12860 Supplier Strem, An Ascensus Company Supplier No. 68680410G
CAS# 10025-75-9. 10g. Erbium(III) chloride hydrate (99.999%-Er) (REO) PURATREM. MFCD00149692. Molecular Weight: 273.64. Molecular Formula: ErCl3∙XH2O. Color/form: pink xtl. Strem# 68-6804. http://www.strem.com/catalog/v/68-6804/