Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) chloride hydrate, REacton™, 99.99% (REO)

Catalog No. AA1130522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1130522 100 g
AA1130514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1130522 Supplier Thermo Scientific Chemicals Supplier No. 01130522
Add to Cart

CAS: 19423-85-9 | Cl3Er | 273.61 g/mol

Erbium(III) chloride hydrate serves as a mild Lewis acid for selective acetalization. It combines with sodium borohydride and used as a selective reducing agent. It is also useful to form enolates from alfa-bromo ketones and oxyallyl cations from alfa,alfa'-dibromo ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-85-9
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.61
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
IUPAC Name erbium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Er+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 273.62 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) chloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-385-0
RTECSNumber KD8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.