Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) chloride, ultra dry, 99.9% (REO)

Catalog No. AA3573906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3573906 5 g
AA3573914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3573906 Supplier Thermo Scientific Chemicals Supplier No. 03573906
Add to Cart

CAS: 10138-41-7 | Cl3Er | 273.609 g/mol

It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-41-7
Molecular Formula Cl3Er
Molecular Weight (g/mol) 273.609
MDL Number MFCD00010989
InChI Key HDGGAKOVUDZYES-UHFFFAOYSA-K
Synonym erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
PubChem CID 66277
IUPAC Name trichloroerbium
SMILES Cl[Er](Cl)Cl

Specifications

Boiling Point 1500°C
Melting Point 774°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF.
Formula Weight 273.62
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) chloride, Ultra dry
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 233-385-0
RTECSNumber KD8575000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.