Learn More
Strem, An Ascensus Company CAS# 10031-51-3. 25g. Erbium(III) nitrate hydrate (99.9%-Er) (REO). MFCD00149691
SDP
Catalog No. 5090116166
Encompass
CAS# 10031-51-3. 25g. Erbium(III) nitrate hydrate (99.9%-Er) (REO). MFCD00149691. Molecular Weight: 353.29. Molecular Formula: Er(NO3)3∙XH2O. Color/form: pink xtl. Strem# 93-6808. http://www.strem.com/catalog/v/93-6808/
Catalog No. 50-901-16166 Supplier Strem, An Ascensus Company Supplier No. 93680825G
CAS# 10031-51-3. 25g. Erbium(III) nitrate hydrate (99.9%-Er) (REO). MFCD00149691. Molecular Weight: 353.29. Molecular Formula: Er(NO3)3∙XH2O. Color/form: pink xtl. Strem# 93-6808. http://www.strem.com/catalog/v/93-6808/