Learn More
Cayman Chemical Erbstatin analog
SDP
Catalog No. 502888574
Encompass_Preferred
A stable, potent analog of erbstatin; inhibits EGFR tyrosine kinase in vitro with an IC50 of 0.14 UG/ml; inhibits EGF-stimulated growth in NIH3T3 cells with an IC50 value of 0.5 UG/ml; delays onset of EGF-induced DNA synthesis
Catalog No. 50-288-8574 Supplier Cayman Chemical Supplier No. 1001023850 MG
Add to Cart
Add to Cart
A stable, potent analog of erbstatin; inhibits EGFR tyrosine kinase in vitro with an IC50 of 0.14 UG/ml; inhibits EGF-stimulated growth in NIH3T3 cells with an IC50 value of 0.5 UG/ml; delays onset of EGF-induced DNA synthesis