Learn More
ERC WIPING PRODUCTS Disposable Polypropylene Beakers, 250 mL, Sterile, Pack of 500
SDP
Catalog No. NC2267258
Encompass_Preferred
a polypropylene microcentrifuge tube rack that holds 0.5mL microcentrifuge tubes. This white rack has 96 tube capacity arranged in an 8x12 matrix. It has alphanumeric coordinates to identify tube positions. The rack's dimensions are 127x86x45mm (5x3.4x1.8in). It can withstand temperatures from -20°C to 121°C (-4°F to 250°F) and is compatible with autoclaving. This product is non-sterile and non-pyrogenic.
This item is not returnable. View return policy
Catalog No. NC2267258 Supplier ERC WIPING PRODUCTS Supplier No. HTNWIMP
a polypropylene microcentrifuge tube rack that holds 0.5mL microcentrifuge tubes. This white rack has 96 tube capacity arranged in an 8x12 matrix. It has alphanumeric coordinates to identify tube positions. The rack's dimensions are 127x86x45mm (5x3.4x1.8in). It can withstand temperatures from -20°C to 121°C (-4°F to 250°F) and is compatible with autoclaving. This product is non-sterile and non-pyrogenic.