Learn More
Aviva Systems Biology Corp ERC1 Antibody - C-terminal region (ARP59491_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59491_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ERC1. Alternate names: Cast2, ELKS, FLJ31750, KIAA1081, MGC12974, RAB6IP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-719