Learn More
ABclonal Technology ERC2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A158101000UL
Encompass_Preferred
This gene encodes a protein that belongs to the Rab3-interacting molecule (RIM)-binding protein family. Members of this protein family form part of the cytomatrix at the active zone (CAZ) complex and function as regulators of neurotransmitter release. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-06662