Learn More
Medchemexpress LLC Ercalcidiol-d3 | 1217467-39-4 | 99.9% | 415.67 | 100 UG
SDP
Catalog No. 5000402080
Encompass_Preferred
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol, a metabolite of Vitamins D2. It serves as an indicator of vitamin D status and can be utilized as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including hydrogen and carbon, are incorporated into this drug molecule. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as an indicator of vitamin D status.
  • Functions as a tracer in quantitative analysis.
  • Serves as an internal standard for NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-02080 Supplier Medchemexpress LLC Supplier No. HY32349S100UG
Add to Cart
Add to Cart
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol, a metabolite of Vitamins D2. It serves as an indicator of vitamin D status and can be utilized as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including hydrogen and carbon, are incorporated into this drug molecule. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as an indicator of vitamin D status.
  • Functions as a tracer in quantitative analysis.
  • Serves as an internal standard for NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.