Learn More
Medchemexpress LLC Ercalcidiol-d3 | 1217467-39-4 | 99.9% | 415.67 | 5 MG
SDP
Catalog No. 5000404629
Encompass_Preferred
Ercalcidiol-d3 is the deuterium-labeled form of Ercalcidiol, which is a metabolite of Vitamin D2. It can be utilized as an indicator of vitamin D status.
  • Use as a tracer.
  • Use as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04629 Supplier Medchemexpress LLC Supplier No. HY32349S5MG
Ercalcidiol-d3 is the deuterium-labeled form of Ercalcidiol, which is a metabolite of Vitamin D2. It can be utilized as an indicator of vitamin D status.
  • Use as a tracer.
  • Use as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.