Learn More
Medchemexpress LLC Ercalcidiol-d3 (25-hydroxy vitamin D2-d3) | 1217467-39-4 | 99.9% | 415.67 | 25 MG
SDP
Catalog No. 5000403363
Encompass_Preferred
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol. Ercalcidiol is a metabolite of Vitamins D2. Ercalcidiol can be used as an indicator of vitamins D status.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-03363 Supplier Medchemexpress LLC Supplier No. 25MGHY32349S
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol. Ercalcidiol is a metabolite of Vitamins D2. Ercalcidiol can be used as an indicator of vitamins D status.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.