Learn More
Medchemexpress LLC Ercalcidiol-d3 (25-Hydroxy vitamin D2-d3) | 1217467-39-4 | 99.9% | 415.67 | 50 MG
SDP
Catalog No. 5000404041
Encompass_Preferred
Ercalcidiol-d3 is a deuterium-labeled version of Ercalcidiol. Ercalcidiol, a metabolite of Vitamin D2, is used as an indicator of vitamin D status. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated, which can impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-04041 Supplier Medchemexpress LLC Supplier No. HY32349S50MG
Ercalcidiol-d3 is a deuterium-labeled version of Ercalcidiol. Ercalcidiol, a metabolite of Vitamin D2, is used as an indicator of vitamin D status. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated, which can impact pharmacokinetic and metabolic profiles of drugs.