Learn More
Medchemexpress LLC Ercalcidiol-d3 (25-Hydroxy Vitamin D2-d3) | 1217467-39-4 | 99.9% | 415.67 | 500 UG
SDP
Catalog No. 5000404622
Encompass_Preferred
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol, a metabolite of Vitamin D2. It can be used as an indicator of vitamin D status. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04622 Supplier Medchemexpress LLC Supplier No. HY32349S500UG
Ercalcidiol-d3 is the deuterium labeled Ercalcidiol, a metabolite of Vitamin D2. It can be used as an indicator of vitamin D status. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.