Learn More
Diagnostic Biosystems ERCC1 ERCC1
Catalog No. 502407387
Encompass_Preferred
This antibody reacts with a 33-36 kDa protein known as ERCC1 (excision repair cross complementing) polypeptide. ERCC1 is required for nucleotide excision repair of damaged DNA and is homologous to RAD10. In mammalian cells, XPG cleaves 3’ of the DNA lesion while ERCC1-XPF complex makes the 5’ incision.
Catalog No. 50-240-7387 Supplier Diagnostic Biosystems Supplier No. PDM151
This antibody reacts with a 33-36 kDa protein known as ERCC1 (excision repair cross complementing) polypeptide. ERCC1 is required for nucleotide excision repair of damaged DNA and is homologous to RAD10. In mammalian cells, XPG cleaves 3’ of the DNA lesion while ERCC1-XPF complex makes the 5’ incision.