Learn More
Aviva Systems Biology Corp ERCC3 Antibody - N-terminal region (ARP37963_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37963_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERCC3. Alternate names: BTF2, GTF2H, RAD25, TFIIH, XPBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62936