Learn More
Aviva Systems Biology Corp ERCC4 Antibody - middle region (ARP58272_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58272_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ERCC4. Alternate names: RAD1, XPF, ERCC11ApplicationWB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-014-62272