Learn More
Aviva Systems Biology Corp ERCC5 Antibody - N-terminal region (ARP54287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERCC5. Alternate names: COFS3, ERCM2, UVDR, XPG, XPGCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61159