Learn More
Aviva Systems Biology Corp ERCC6L Antibody - N-terminal region (ARP56997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC6L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERCC6L. Alternate names: FLJ20105, MGC131695, PICHApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53956